Sodium pivalate hydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethylacetic acid sodium salt hydrate, Sodium trimethylacetate hydrate, Propanoic acid, 2,2-dimethyl-, sodium salt, hydrate

36471-0100 36471-0500
TS36471-0100EA 76 USD
TS36471-0100 TS36471-0500
Sodium pivalate hydrate 99%
Sodium pivalate hydrate
Formula: C₅H₁₁NaO₃
MW: 142.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150782
CAS Number: 143174-36-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤7 % (K.F.)
Titration with HClO4 ≥98.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR