2,2,3,3-Tetramethoxybutane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Butane, 2,2,3,3-tetramethoxy-
45093-0050
45093-0250
TS45093-0050EA
110
USD
TS45093-0050
TS45093-0250
2,2,3,3-Tetramethoxybutane 95%
2,2,3,3-Tetramethoxybutane
Formula:
C₈H₁₈O₄ MW: 178.23 g/mol Boiling Pt: 172…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD25458126 CAS Number: 176798-33-7 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...