2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Oxiran-2-ylmethyl)isoindoline-1,3-dione, N-(2,3-Epoxypropyl)phthalimide

17004-0100 17004-0250
TS17004-0100EA 126 USD
TS17004-0100 TS17004-0250
2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione 95%
2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione
Formula: C₁₁H₉NO₃
MW: 203.2 g/mol
Melting Pt: 94…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00005896
CAS Number: 5455-98-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 92 to 102 °C
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR