tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-BOC-L-Valinol, Boc-Valinol, (S)-tert-butyl (1-hydroxy-3-methylbutan-2-yl)carbamate

39923-0010 39923-0050
TS39923-0010EA 54 USD
TS39923-0010 TS39923-0050
tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate 96%
tert-Butyl (1S)-1-(hydroxymethyl)-2-methylpropylcarbamate
Formula: C₁₀H₂₁NO₃
MW: 203.28 g/mol
Boiling Pt: 218 °C (760 mmHg)
Density: 0.995 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00082635
CAS Number: 79069-14-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥95 %
Specific optical rotation -21° to -25° (20°C, 589 nm) (c=1%, Chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR