Methyl-4-aminobutanoate hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: H-γ-Abu-OMe.HCl

44291-0250 44291-1000
TS44291-0250EA 140 USD
TS44291-0250 TS44291-1000
Methyl-4-aminobutanoate hydrochloride 99%
Methyl-4-aminobutanoate hydrochloride
Formula: C₅H₁₁NO₂·HCl
MW: 153.61 g/mol
Melting Pt: 118…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00043270
CAS Number: 13031-60-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric 98.5 to 101.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR