3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-tert-Butyl-1-methylpyrazole-5-carboxylic Acid

45262-0050 45262-0500
TS45262-0050EA 222 USD
TS45262-0050 TS45262-0500
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid 95%
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid
Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00173812
CAS Number: 175277-11-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 150 to 156 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR