Thapsigargin 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-Acetoxy-4-(butyryloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-(((Z)-2-methylbut-2-enoyl)oxy)-2-oxo-2,3,3a,4,5,6,6a,7,8,9b-decahydroazuleno[4,5-b]furan-7-yl octanoate

32857-0010 32857-0050
TS32857-0010EA 161 USD
TS32857-0010 TS32857-0050
Thapsigargin 97%
Thapsigargin
Formula: C₃₄H₅₀O₁₂
MW: 650.762 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00083511
CAS Number: 67526-95-8

Order Now

Specification Test Results

Appearance Colorless Film
HPLC ≥96.0 %
TLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR