Isopropyl methanesulphonate 99%

Supplier: Thermo Scientific Chemicals
Danger

41284-0050 41284-0250 41284-1000
TS41284-0050EA 119 USD
TS41284-0050 TS41284-0250 TS41284-1000
Isopropyl methanesulphonate 99%
Isopropyl methanesulfonate
Formula: C₄H₁₀O₃S
MW: 138.181 g/mol
Boiling Pt: 82 °C (6 mmHg)
Density: 1.145 g/cm³
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047802
CAS Number: 926-06-7
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4210 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR