Pivalic acid hydrazide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pivalohydrazide, Pivaloyl hydrazide

37775-0010 37775-0050
TS37775-0010EA 58 USD
TS37775-0010 TS37775-0050
Pivalic acid hydrazide 98+%
Pivalic acid hydrazide

Appearance: White Crystals

Formula: C₅H₁₂N₂O
MW: 116.164 g/mol
Boiling Pt: 95…110 °C (1mbar)
Melting Pt: 65…67 °C
Storage Temperature: Ambient
MDL Number: MFCD01310839
CAS Number: 42826-42-6
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White Crystals
Infrared spectrum Conforms
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR