Methyl-2-(m-tolyl)acetate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 3-methylphenylacetate

37408-0050 37408-0250
TS37408-0050EA 53 USD
TS37408-0050 TS37408-0250
Methyl-2-(m-tolyl)acetate 98%
Methyl-2-(m-tolyl)acetate

Appearance: Clear colorless to light yellow Liquid

Formula: C₁₀H₁₂O₂
MW: 164.204 g/mol
Boiling Pt: 117 °C (13 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00968468
CAS Number: 53088-69-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5045 to 1.5065 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR