methyl cyclobutanecarboxylate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl cyclobutanecarboxylate

36554-0050 36554-0250
TS36554-0050EA 97 USD
TS36554-0050 TS36554-0250
methyl cyclobutanecarboxylate 98%
methyl cyclobutanecarboxylate
Formula: C₆H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00095142
CAS Number: 765-85-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4300 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR