2,2-Di(tert-butylperoxy)butane 50% in aromatic free mineral spirit

Supplier: Thermo Scientific Chemicals
Danger

34983-0100 34983-5000
TS34983-0100EA 24 USD
TS34983-0100 TS34983-5000
2,2-Di(tert-butylperoxy)butane 50% in aromatic free mineral spirit
2,2-Di(tert-butylperoxy)butane

Appearance: Clear solution

Formula: C₁₂H₂₆O₄
Density: 0.805 g/cm³
Flash Pt: 43 °C
Storage Temperature: Refrigerator
CAS Number: 2167-23-9
UN: 3103
ADR: 5.2,

Order Now

Specification Test Results

Appearance Clear solution
Titration Iodometric 49 to 51 %
Color scale ≤10 APHA
Impurity ≤0.3 % Hydroperoxides (as TBHP)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR