Diethyl (3,3-diethoxypropyl)phosphonate 95% stabilised

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with sodium carbonate (Na₂CO₃) max. 1% w/w

34693-0050 34693-0250 34693-1000
TS34693-0050EA 64 USD
TS34693-0050 TS34693-0250 TS34693-1000
Diethyl (3,3-diethoxypropyl)phosphonate 95% stabilised
Diethyl (3,3-diethoxypropyl)phosphonate
Formula: C₁₁H₂₅O₅P
Storage Temperature: Ambient
MDL Number: MFCD00800259
CAS Number: 15110-17-5

Order Now

Specification Test Results

Appearance Clear light yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4320 (20°C, 589 nm)
GC ≥94 %
Stabilizer ≤1 % Na2CO3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR