3-Pyrazolidinone hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyrazolidin-3-one hydrochloride
33549-0010
33549-0050
33549-0250
TS33549-0010EA
75
USD
TS33549-0010
TS33549-0050
TS33549-0250
3-Pyrazolidinone hydrochloride 98%
3-Pyrazolidinone hydrochloride
Formula:
C₃H₇ClN₂O MW: 122.552 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08594075 CAS Number: 1752-88-1 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...