Diethylphosphonoacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Diethoxyphosphoryl)acetic acid, Diethyl (hydroxycarbonylmethyl)phosphonate

33067-0050 33067-0250
TS33067-0050EA 29 USD
TS33067-0050 TS33067-0250
Diethylphosphonoacetic acid 98%
Diethylphosphonoacetic acid

Appearance: Clear colorless to yellow Viscous liquid

Formula: C₆H₁₃O₅P
MW: 196.139 g/mol
Boiling Pt: 150 °C
Density: 1.22 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00192032
CAS Number: 3095-95-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR