Methyl (2E)-pent-2-enoate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl trans-2-pentenoate, (E)-Methyl pent-2-enoate

32972-0050 32972-0250
TS32972-0050EA 107 USD
TS32972-0050 TS32972-0250
Methyl (2E)-pent-2-enoate 95%
Methyl (2E)-pent-2-enoate

Appearance: Clear colorless Liquid

Formula: C₆H₁₀O₂
MW: 114.144 g/mol
Flash Pt: 20 °C
Storage Temperature: Freezer
MDL Number: MFCD00137611
CAS Number: 15790-88-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4320 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR