1-(4-Chlorophenyl)ethanamine 90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(4-Chlorophenyl)ethylamine

32947-0010 32947-0050
TS32947-0010EA 47 USD
TS32947-0010 TS32947-0050
1-(4-Chlorophenyl)ethanamine 90%
1-(4-Chlorophenyl)ethanamine

Appearance: Clear yellow Liquid

Formula: C₈H₁₀ClN
MW: 155.625 g/mol
Boiling Pt: 232 °C (760 mmHg)
Density: 1.1 g/cm³
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00044766
CAS Number: 6299-02-1
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5455 to 1.5475 (20°C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR