Methyl-1-hydroxy-1-cyclopropane carboxylate 90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 1-hydroxycyclopropanecarboxylate

30211-0010 30211-0050
TS30211-0010EA 200 USD
TS30211-0010 TS30211-0050
Methyl-1-hydroxy-1-cyclopropane carboxylate 90%
Methyl-1-hydroxy-1-cyclopropane carboxylate

Appearance: Clear yellow Liquid

Formula: C₅H₈O₃
MW: 116.116 g/mol
Boiling Pt: 70 °C (13 mmHg)
Flash Pt: 17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093884
CAS Number: 33689-29-1
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR