2-(3-Methoxyallylidene)malonic acid dimethyl ester 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(3-Methoxyallylidene)malonic acid dimethyl ester

30209-0025 30209-0100
TS30209-0025EA 82 USD
TS30209-0025 TS30209-0100
2-(3-Methoxyallylidene)malonic acid dimethyl ester 95%
2-(3-Methoxyallylidene)malonic acid dimethyl ester
Formula: C₉H₁₂O₅
MW: 200.19 g/mol
Boiling Pt: 120…125 °C (.3 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00800589
CAS Number: 41530-32-9

Order Now

Specification Test Results

Appearance Clear yellow-green Low melting solid
Infrared spectrum Conforms
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR