(2-Aminoethyl)phosphonic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-AEP, 2-Aminoethylphosphonic acid

29163-0010 29163-2500
TS29163-0010EA 174 USD
TS29163-0010 TS29163-2500
(2-Aminoethyl)phosphonic acid 99%
(2-Aminoethyl)phosphonic acid
Formula: C₂H₈NO₃P
MW: 125.064 g/mol
Melting Pt: 296 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008182
CAS Number: 2041-14-7

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR