Methyl-1-cyclohexene-1-carboxylate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl cyclohex-1-enecarboxylate

21371-0050 21371-0250
TS21371-0050EA 134 USD
TS21371-0050 TS21371-0250
Methyl-1-cyclohexene-1-carboxylate 98%
Methyl-1-cyclohexene-1-carboxylate

Appearance: Clear colorless Liquid

Formula: C₈H₁₂O₂
MW: 140.182 g/mol
Boiling Pt: 190…192 °C (760 mmHg)
Density: 1.03 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00001544
CAS Number: 18448-47-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4790 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR