Ethyl-3-methyl-2-oxobutyrate 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl 3-methyl-2-oxobutanoate

20380-0050 20380-0250
TS20380-0050EA 64 USD
TS20380-0050 TS20380-0250
Ethyl-3-methyl-2-oxobutyrate 95%
Ethyl-3-methyl-2-oxobutyrate

Appearance: Clear light yellow Liquid

Formula: C₇H₁₂O₃
Boiling Pt: 62 °C (11 mmHg)
Density: 0.98 g/cm³
Flash Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009122
CAS Number: 20201-24-5
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4130 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR