Diethyl-bis(hydroxymethyl)malonate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 2,2-bis(hydroxymethyl)malonate

18953-0250 18953-1000 18953-5000
TS18953-0250EA 75 USD
TS18953-0250 TS18953-1000 TS18953-5000
Diethyl-bis(hydroxymethyl)malonate 95%
Diethyl-bis(hydroxymethyl)malonate

Appearance: White to off-white Crystalline solid

Formula: C₉H₁₆O₆
MW: 220.221 g/mol
Melting Pt: 45…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009130
CAS Number: 20605-01-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Melting point 45 to 52 °C
GC ≥94 %
Titration with HClO4 97.0 to 102.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR