6-Hydroxyhexanoic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Hydroxycaproic acid

34224-0010 34224-0050 34224-0250
TS34224-0010EA 176 USD
TS34224-0010 TS34224-0050 TS34224-0250
6-Hydroxyhexanoic acid 95%
6-Hydroxyhexanoic acid
Formula: C₆H₁₂O₃
MW: 132.159 g/mol
Boiling Pt: 113…116 °C (760 mmHg)
Melting Pt: 38…40 °C
Density: 0.981 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00046560
CAS Number: 1191-25-9

Order Now

Specification Test Results

Appearance White Low melting crystalline solid
Infrared spectrum Conforms
Additional info may contain variable amounts of water
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR