(1S,4R)-(+)-4-Acetoxy-2-cyclopentene-1-ol 99% stabilized

Supplier: Thermo Scientific Chemicals

Synonyms: (1S,4R)-cis-4-Acetoxy-2-cyclopenten-1-ol, (1R,4S)-4-hydroxycyclopent-2-enyl acetate, (1R,3S)-(+)-4-Cyclopentene-1,3-diol 1-acetate

39934-0010 39934-0050
TS39934-0010EA 230 USD
TS39934-0010 TS39934-0050
(1S,4R)-(+)-4-Acetoxy-2-cyclopentene-1-ol 99% stabilized
(1S,4R)-(+)-4-Acetoxy-2-cyclopentene-1-ol
Formula: C₇H₁₀O₃
MW: 142.154 g/mol
Melting Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078933
CAS Number: 60410-16-4

Order Now

Specification Test Results

Appearance White to light yellow Solid
Infrared spectrum Conforms
Melting point 49 to 53 °C
GC ≥98.5 %
Specific optical rotation +67° to +69° (20 °C, 589 nm) (c=2.3, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR