DL-3-Phenyllactic acid 97%

Supplier: Thermo Scientific Chemicals

Synonyms: DL-β-Phenyllactic acid

42777-0010 42777-0100
TS42777-0010EA 29 USD
TS42777-0010 TS42777-0100
DL-3-Phenyllactic acid 97%
DL-3-Phenyllactic acid

Appearance: White to off-white Crystalline powder

Formula: C₉H₁₀O₃
MW: 166.176 g/mol
Melting Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065928
CAS Number: 828-01-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 94 to 98 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR