tert-Butyl-N-allylcarbamate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Boc-allylamine, N-(tert-Butoxycarbonyl)allylamine, N-Allylcarbamic Acid tert-Butyl Ester, tert-Butyl allylcarbamate

43477-0100 43477-0500
TS43477-0100EA 77 USD
TS43477-0100 TS43477-0500
tert-Butyl-N-allylcarbamate 98%
tert-Butyl-N-allylcarbamate
Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 48…50 °C (1 mmHg)
Melting Pt: 34.9…38.3 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00191870
CAS Number: 78888-18-3

Order Now

Specification Test Results

Appearance Colorless to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 33 to 40 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR