Sodium-DL-3-hydroxybutyrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium 3-hydroxybutanoate, DL-3-Hydroxybutyric acid Sodium salt

21501-0250 21501-1000
TS21501-0250EA 117 USD
TS21501-0250 TS21501-1000
Sodium-DL-3-hydroxybutyrate 99%
Sodium-DL-3-hydroxybutyrate
Formula: C₄H₈O₃
MW: 126.09 g/mol
Melting Pt: 161…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016716
CAS Number: 150-83-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 161 to 167 °C (Dec)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR