Sodium benzenesulfonate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Benzenesulphonic acid Sodium Salt, Benzenesulphonic acid, sodium salt, Benzenesulphonate sodium
40185-0250
40185-1000
40185-5000
TS40185-0250EA
24
USD
TS40185-0250
TS40185-1000
TS40185-5000
Sodium benzenesulfonate 98%
Sodium benzenesulfonate
Formula:
C₆H₆O₃S Melting Pt: 450 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065179 CAS Number: 515-42-4 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (1 g, 105°C, 3 h) |
Solubility | (5% in water) Clear colorless to almost colorless |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...