Sodium-4-hydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxybenzoate sodium, Sodium 4-hydroxybenzoate, p-Hydroxybenzoic acid Monosodium Salt, 4-Hydroxybenzoic Acid Sodium Salt

39152-0250 39152-1000
TS39152-0250EA 63 USD
TS39152-0250 TS39152-1000
Sodium-4-hydroxybenzoate 98%
Sodium-4-hydroxybenzoate

Appearance: White to light beige Crystalline powder

Formula: C₇H₆O₃
MW: 160.10 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00016530
CAS Number: 114-63-6

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR