N-Benzoyl-N-phenylhydroxylamine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Tantalon, N-Phenylbenzohydroxamic Acid, N-Hydroxy-N-phenylbenzamide

41737-0050 41737-0250 41737-1000
TS41737-0050EA 93 USD
TS41737-0050 TS41737-0250 TS41737-1000
N-Benzoyl-N-phenylhydroxylamine 98%
N-Benzoyl-N-phenylhydroxylamine
Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Melting Pt: 119…123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002111
CAS Number: 304-88-1

Order Now

Specification Test Results

Appearance Light beige Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 119 to 123 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR