N-(tert-Butoxycarbonyl)-1,4-diaminobutane 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-(4-Aminobutyl)carbamic Acid tert-Butyl Ester, N-(tert-Butoxycarbonyl)-1,4-butanediamine, NH2-C4-NH-Boc, N-Boc-1,4-diaminobutane, N-Boc-1,4-butanediamine, tert-Butyl N-(4-Aminobutyl)carbamate, tert-Butyl (4-aminobutyl)carbamate

42619-0010 42619-0050
TS42619-0010EA 106 USD
TS42619-0010 TS42619-0050
N-(tert-Butoxycarbonyl)-1,4-diaminobutane 95%
N-(tert-Butoxycarbonyl)-1,4-diaminobutane

Appearance: Clear colorless to light yellow Liquid

Formula: C₉H₂₀N₂O₂
MW: 188.27 g/mol
Density: 0.984 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00210019
CAS Number: 68076-36-8
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94 %
NMR consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR