Methyl-2-(bromomethyl)acrylate 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Bromomethyl)acrylic Acid Methyl Ester, Methyl 2-(bromomethyl)acrylate

36763-0010 36763-0050 36763-0250
TS36763-0010EA 141 USD
TS36763-0010 TS36763-0050 TS36763-0250
Methyl-2-(bromomethyl)acrylate 96%
Methyl-2-(bromomethyl)acrylate

Appearance: Clear colorless to light brown Liquid

Formula: C₅H₇BrO₂
MW: 179.01 g/mol
Boiling Pt: 35…37 °C (13 mmHg)
Density: 1.489 g/cm³
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011697
CAS Number: 4224-69-5
UN: 3334
ADR: 9 III

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4910 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR