Formaldehyde sodium bisulphite 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Sodium Hydroxymethanesulphonate, Hydroxymethanesulphonic acid sodium salt

15069-0010 15069-0050 15069-0051
TS15069-0010EA 59 USD
TS15069-0010 TS15069-0050 TS15069-0051
Formaldehyde sodium bisulphite 95%
Formaldehyde sodium bisulfite

Appearance: White to off-white Powder

Formula: CH₄O₄S
MW: 134.08 g/mol
Melting Pt: 200 °C
Flash Pt: 184 °C
Storage Temperature: Ambient
MDL Number: MFCD00044664
CAS Number: 870-72-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration ≥94 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR