Dibutyl phosphate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dibutylphosphate, Phosphoric Acid Dibutyl Ester

40664-0025 40664-2500 40664-5000
TS40664-0025EA 274 USD
TS40664-0025 TS40664-2500 TS40664-5000
Dibutyl phosphate 97%
Dibutyl phosphate

Appearance: Clear liquid

Formula: C₈H₁₉O₄P
MW: 210.21 g/mol
Boiling Pt: 250 °C (760 mmHg)
Melting Pt: –13 °C
Density: 1.058 g/cm³
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00015245
CAS Number: 107-66-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4290 (20°C, 589 nm)
Water ≤0.3 %
Color scale ≤100 APHA
Titration with NaOH ≥96.0 %
Impurity ≤3.0 % Tributyl phosphate
Acid value 259 to 267 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR