cis-1,2,3,6-Tetrahydrophthalimide 96%

Supplier: Thermo Scientific Chemicals
Warning

13823-0050 13823-1000 13823-2500
TS13823-0050EA 19 USD
TS13823-0050 TS13823-1000 TS13823-2500
cis-1,2,3,6-Tetrahydrophthalimide 96%
cis-1,2,3,6-Tetrahydrophthalimide
Formula: C₈H₉NO₂
Melting Pt: 131…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00005880
CAS Number: 1469-48-3

Order Now

Specification Test Results

Appearance Yellow to light beige to light brown Powder or flakes
Infrared spectrum Conforms
Melting point 131 to 138 °C
Titration with NaOH ≥95 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR