5-Vinylbicyclo[2.2.1]hept-2-ene 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Vinyl-2-norbornene

16486-0025 16486-0250 16486-5000
TS16486-0025EA 453 USD
TS16486-0025 TS16486-0250 TS16486-5000
5-Vinylbicyclo[2.2.1]hept-2-ene 98% stabilized
5-Vinylbicyclo[2.2.1]hept-2-ene
Formula: C₉H₁₂
MW: 120.20 g/mol
Boiling Pt: 141 °C (760 mmHg)
Melting Pt: –80 °C
Flash Pt: 27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080691
CAS Number: 3048-64-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4820 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤50 APHA
Stabilizer 100 to 200 ppm t-Butylcatechol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR