4-Acryloylmorpholine 97%

Supplier: Thermo Scientific Chemicals
Danger

43925-0500 43925-2500
TS43925-0500EA 53 USD
TS43925-0500 TS43925-2500
4-Acryloylmorpholine 97%
4-Acryloylmorpholine
Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 158 °C (50 mmHg)
Melting Pt: –35 °C
Density: 1.122 g/cm³
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047413
CAS Number: 5117-12-4

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Water ≤2.50 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR