3-Hydroxy-2,2,4-trimethylpentyl isobutyrate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2,4-Trimethyl-1,3-pentanediol monoisobutyrate, 2,2,4-Trimethyl-1,3-pentanediol 1-monoisobutyrate, Isobutyric Acid 3-Hydroxy-2,2,4-trimethylpentyl Ester, 2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate)

42201-0025 42201-5000
TS42201-0025EA 96 USD
TS42201-0025 TS42201-5000
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate 98+%
3-Hydroxy-2,2,4-trimethylpentyl isobutyrate
Formula: C₁₂H₂₄O₃
Boiling Pt: 244 °C (760 mmHg)
Density: 0.95 g/cm³
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00148967
CAS Number: 25265-77-4

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4430 (20°C, 589 nm)
GC ≥98.0 % (sum two isomers)
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR