3-(tert-Butoxycarbonylamino)-1-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Boc-amino)-1-propanol, N-(3-Hydroxypropyl)carbamic Acid tert-Butyl Ester, Boc-β-Ala-ol, 3-(BOC-amino)-1-propanol, tert-Butyl N-(3-Hydroxypropyl)carbamate

39980-0100 39980-0500
TS39980-0100EA 69 USD
TS39980-0100 TS39980-0500
3-(tert-Butoxycarbonylamino)-1-propanol 97%
3-(tert-Butoxycarbonylamino)-1-propanol
Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
Density: 1.025 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00191883
CAS Number: 58885-58-8

Order Now

Specification Test Results

Appearance Colorless to yellow to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR