2-Sulphobenzaldehyde sodium salt 90% tech.

Supplier: Thermo Scientific Chemicals

21932-0025 21932-5000
TS21932-0025EA 545 USD
TS21932-0025 TS21932-5000
2-Sulphobenzaldehyde sodium salt 90% tech.
2-Sulfobenzaldehyde sodium salt
Formula: C₇H₆O₄S
Storage Temperature: Ambient
MDL Number: MFCD00007478
CAS Number: 1008-72-6

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥88 %
Sodium chloride (NaCl) ≤3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR