2-Ethylhexyl-4-methoxycinnamate 98% stabilized

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methoxycinnamic Acid 2-Ethylhexyl Ester, 4-Methoxycinnamic Acid Octyl Ester, Octinoxate, Octyl 4-Methoxycinnamate, 2-Ethylhexyl 3-(4-methoxyphenyl)acrylate

29116-0250 29116-1000
TS29116-0250EA 81 USD
TS29116-0250 TS29116-1000
2-Ethylhexyl-4-methoxycinnamate 98% stabilized
2-Ethylhexyl-4-methoxycinnamate
Formula: C₁₈H₂₆O₃
MW: 290.40 g/mol
Density: 1.009 g/cm³
Flash Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00072582
CAS Number: 5466-77-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5430 to 1.5470 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.05 to 0.1 % BHT
Specific gravity (25/25°C) 1.007 to 1.012
Specific absorbance A (1%/1cm) ≥830 (at 307 to 308 nm, MeOH)
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR