2-(tert-Butoxycarbonylamino)-1-ethanol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonyl)ethanolamine, N-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(Boc-amino)-1-ethanol, Boc-Glycinol

30317-0050 30317-0250
TS30317-0050EA 61 USD
TS30317-0050 TS30317-0250
2-(tert-Butoxycarbonylamino)-1-ethanol 98%
2-(tert-Butoxycarbonylamino)-1-ethanol

Appearance: Clear colorless to light yellow Viscous liquid

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Density: 1.042 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00056657
CAS Number: 26690-80-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4485 to 1.4505 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR