2-(Carbobenzoxyamino)-1-ethanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
43395-0050
43395-0250
TS43395-0050EA
36
USD
TS43395-0050
TS43395-0250
2-(Carbobenzoxyamino)-1-ethanol 98%
2-(Carbobenzoxyamino)-1-ethanol
Formula:
C₁₀H₁₃NO₃ Boiling Pt: 170 °C (.5Torr) Melting Pt: 61…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191060 CAS Number: 77987-49-6 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 60 to 65 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...