1-Adamantaneacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Adamantan-1-yl)acetic acid, 1-Carboxymethyladamantane

33156-0010 33156-0050 33156-0100
TS33156-0010EA 36 USD
TS33156-0010 TS33156-0050 TS33156-0100
1-Adamantaneacetic acid 98%
1-Adamantaneacetic acid
Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00074728
CAS Number: 4942-47-6

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 134 to 138 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR