1-Acetamidoadamantane 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-1-Adamantylacetamide, N-Acetyl-1-adamantanamine

13360-0250 13360-1000
TS13360-0250EA 52 USD
TS13360-0250 TS13360-1000
1-Acetamidoadamantane 99+%
1-Acetamidoadamantane

Appearance: White to yellow Crystalline powder

Formula: C₁₂H₁₉NO
MW: 193.29 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00074730
CAS Number: 880-52-4

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 149 °C
GC ≥99.0 %
Elemental Analysis 52.4 to 56.8 % C
Specific optical rotation -40° to -50° (20 °C, 589 nm) (c=1 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR