1,3,2-Dioxathiolane-2,2-dioxide 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,2-Dioxathiolane 2,2-dioxide, Ethylene Sulphate

42663-0010 42663-0050
TS42663-0010EA 31 USD
TS42663-0010 TS42663-0050
1,3,2-Dioxathiolane-2,2-dioxide 96%
1,3,2-Dioxathiolane-2,2-dioxide

Appearance: White to beige Powder

Formula: C₂H₄O₄S
MW: 124.11 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00221769
CAS Number: 1072-53-3

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 95 to 99 °C
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR