(R)-Glycidyl-3-nitrobenzenesulphonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-(-)-Glycidyl nosylate, 3-Nitrobenzenesulphonic acid (R)-Glycidyl Ester, (R)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate

29641-0025 29641-0100
TS29641-0025EA 63 USD
TS29641-0025 TS29641-0100
(R)-Glycidyl-3-nitrobenzenesulphonate 98%
(R)-Glycidyl-3-nitrobenzenesulfonate
Formula: C₉H₉NO₆S
MW: 259.23 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013436
CAS Number: 115314-17-5

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 59 to 63 °C
GC ≥97.5 %
Specific optical rotation (20 °C, 589 nm) (c=2, CHCl3) < -21.5°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR