(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-tert-Butyl pyrrolidin-3-ylcarbamate, (3S)-(-)-3-(Boc-amino)pyrrolidine
43790-0010
43790-0050
TS43790-0010EA
74
USD
TS43790-0010
TS43790-0050
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 99%
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine
Formula:
C₉H₁₈N₂O₂ MW: 186.26 g/mol Melting Pt: 77.8…80.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143194 CAS Number: 122536-76-9 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 72 to 82 °C |
GC | ≥98.5 % |
Specific optical rotation | -17° to -23° (20 °C, 589 nm) (c=1, ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...