(+)-Diacetyl-L-tartaric anhydride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3R,4R)-2,5-Dioxotetrahydrofuran-3,4-diyl diacetate

33604-0050 33604-0250 33604-1000
TS33604-0050EA 28 USD
TS33604-0050 TS33604-0250 TS33604-1000
(+)-Diacetyl-L-tartaric anhydride 97%
(+)-Diacetyl-L-tartaric anhydride
Formula: C₈H₈O₇
MW: 216.15 g/mol
Melting Pt: 130…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037918
CAS Number: 6283-74-5

Order Now

Specification Test Results

Appearance White Powder or crystals or needles
Infrared spectrum Conforms
Melting point 130 to 135 °C
Specific optical rotation +56° to +62° (20 °C, 589 nm) (c=6, acetone)
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR